MAT196005755 Matrix Scientific

N-1H-Imidazol-2-ylacetamide

  • MDL number MFCD09455238
  • Cas Number : 52737-49-2
  • Molecular Weight : 125.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/036984.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/036984.html
Availability details for
Quantity
  • Availability
  • Add to Cart