MAT196005762 Matrix Scientific

N-[(5-Methoxy-1H-indol-3-yl)methyl]-N-(pyridin-3-ylmethyl)amine

  • MDL number MFCD11982981
  • Cas Number : 1114597-59-9
  • Molecular Weight : 267.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/036991.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/036991.html
Availability details for
Quantity
  • Availability
  • Add to Cart