MAT196005799 Matrix Scientific

4-Chloro-2-methyl-6-propylpyrimidine

  • MDL number MFCD07365353
  • Cas Number : 89967-20-4
  • Molecular Weight : 170.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/037028.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/037028.html
Availability details for
Quantity
  • Availability
  • Add to Cart