MAT196005801 Matrix Scientific

(2Z)-{2-[(Chloroacetyl)amino]-1,3-thiazol-4-yl}(methoxyimino)acetic acid

  • MDL number MFCD00067025
  • Cas Number : 64486-18-6
  • Molecular Weight : 277.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/037030.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/037030.html
Availability details for
Quantity
  • Availability
  • Add to Cart