MAT196005828 Matrix Scientific

2-Bromo-4-(1,3-dioxo-1,3-dihydro-2H-isoindol-2-yl)butanoic acid

  • MDL number MFCD12027048
  • Cas Number : 35197-64-9
  • Molecular Weight : 312.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/037057.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/037057.html
Availability details for
Quantity
  • Availability
  • Add to Cart