MAT196005849 Matrix Scientific

3-(2-Hydroxy-1-naphthyl)propanenitrile

  • MDL number MFCD10037626
  • Cas Number : 14233-73-9
  • Molecular Weight : 197.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/037078.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/037078.html
Availability details for
Quantity
  • Availability
  • Add to Cart