MAT196005869 Matrix Scientific

4-(Piperazin-1-ylsulfonyl)aniline

  • MDL number MFCD03444527
  • Cas Number : 69249-13-4
  • Molecular Weight : 241.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/037098.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/037098.html
Availability details for
Quantity
  • Availability
  • Add to Cart