MAT196005882 Matrix Scientific

1-(3-Ethoxyphenyl)methanamine

  • MDL number MFCD03410991
  • Cas Number : 93071-76-2
  • Molecular Weight : 151.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/037111.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/037111.html
Availability details for
Quantity
  • Availability
  • Add to Cart