MAT196005928 Matrix Scientific

5-Hydroxypyrazine-2-carboxylic acid

  • MDL number MFCD00190599
  • Cas Number : 34604-60-9
  • Molecular Weight : 140.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/037157.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/037157.html
Availability details for
Quantity
  • Availability
  • Add to Cart