MAT196005932 Matrix Scientific

(3-Hydroxy-1H-pyrazol-4-yl)acetic acid

  • MDL number MFCD09837445
  • Cas Number : 876716-99-3
  • Molecular Weight : 142.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/037161.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/037161.html
Availability details for
Quantity
  • Availability
  • Add to Cart