MAT196005944 Matrix Scientific

Ethyl 2,5-dichloronicotinate

  • MDL number MFCD07636850
  • Cas Number : 148065-10-5
  • Molecular Weight : 220.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/037173.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/037173.html
Availability details for
Quantity
  • Availability
  • Add to Cart