MAT196005984 Matrix Scientific

2-Phenyl-1-benzofuran-5-amine

  • MDL number MFCD04971060
  • Cas Number : 77084-15-2
  • Molecular Weight : 209.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/037213.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/037213.html
Availability details for
Quantity
  • Availability
  • Add to Cart