MAT196005989 Matrix Scientific

6-Methyl-4-oxo-4H-pyrido[1,2-a]pyrimidine-3-carboxylic acid

  • MDL number MFCD01690160
  • Cas Number : 32092-27-6
  • Molecular Weight : 204.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/037218.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/037218.html
Availability details for
Quantity
  • Availability
  • Add to Cart