MAT196006030 Matrix Scientific

2-Methyl-2-pyrrolidin-1-ylpropanoic acidhydrochloride

  • MDL number MFCD12027087
  • Cas Number : 898403-15-1
  • Molecular Weight : 193.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/037248.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/037248.html
Availability details for
Quantity
  • Availability
  • Add to Cart