MAT196006044 Matrix Scientific

2-[1-(Methylsulfonyl)piperidin-3-yl]ethanaminehydrochloride

  • MDL number MFCD12027120
  • Cas Number : 1185302-17-3
  • Molecular Weight : 242.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/037361.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/037361.html
Availability details for
Quantity
  • Availability
  • Add to Cart