MAT196006047 Matrix Scientific

(4,5-Dimethyl-1H-benzimidazol-2-yl)methylaminehydrochloride

  • MDL number MFCD12027127
  • Cas Number : 1185302-37-7
  • Molecular Weight : 211.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/037394.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/037394.html
Availability details for
Quantity
  • Availability
  • Add to Cart