MAT196006055 Matrix Scientific

Ethyl 3-pyrrolidin-1-ylpropanoate

  • MDL number MFCD00020861
  • Cas Number : 6317-35-7
  • Molecular Weight : 171.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/037261.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/037261.html
Availability details for
Quantity
  • Availability
  • Add to Cart