MAT196006072 Matrix Scientific

2-Methyl-4-piperazin-1-yl-6-(trifluoromethyl)-pyrimidine

  • MDL number MFCD07365354
  • Cas Number : 893752-50-6
  • Molecular Weight : 246.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/037278.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/037278.html
Availability details for
Quantity
  • Availability
  • Add to Cart