MAT196006093 Matrix Scientific

[1-(2-Furylmethyl)piperidin-4-yl]methylamine

  • MDL number MFCD09837352
  • Cas Number : 725211-89-2
  • Molecular Weight : 194.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient/argon
Country of Origin USA
Product Link http://www.matrixscientific.com/037299.html

Safety Information

hcodes Irritant/Stored under Argon
MSDS http://www.matrixscientific.com/037299.html
Availability details for
Quantity
  • Availability
  • Add to Cart