MAT196006102 Matrix Scientific

(3-Chlorophenyl)(4-piperazin-1-ylphenyl)methanone

  • MDL number MFCD06740192
  • Cas Number : 886506-39-4
  • Molecular Weight : 300.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/037308.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/037308.html
Availability details for
Quantity
  • Availability
  • Add to Cart