MAT196006132 Matrix Scientific

2-(4-Methyl-1H-pyrazol-1-yl)propanoic acid

  • MDL number MFCD03419650
  • Cas Number : 1005582-20-6
  • Molecular Weight : 154.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/037338.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/037338.html
Availability details for
Quantity
  • Availability
  • Add to Cart