MAT196006142 Matrix Scientific

2-Cyano-N-isopropylacetamide

  • MDL number MFCD00828755
  • Cas Number : 52573-74-7
  • Molecular Weight : 126.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/037348.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/037348.html
Availability details for
Quantity
  • Availability
  • Add to Cart