MAT196006173 Matrix Scientific

5-(3-Fluorophenyl)-1,3,4-oxadiazol-2-amine

  • MDL number MFCD09028454
  • Cas Number : 341978-66-3
  • Molecular Weight : 179.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/037379.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/037379.html
Availability details for
Quantity
  • Availability
  • Add to Cart