MAT196006215 Matrix Scientific

3-(3-Methylphenyl)isoxazol-5-amine

  • MDL number MFCD06199358
  • Cas Number : 86685-97-4
  • Molecular Weight : 174.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/037421.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/037421.html
Availability details for
Quantity
  • Availability
  • Add to Cart