MAT196006221 Matrix Scientific

2,2,3,3,4,4,5,5-Octafluoropentanal

  • MDL number MFCD00155897
  • Cas Number : 2648-47-7
  • Molecular Weight : 230.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient/argon
Country of Origin USA
Boiling point 83-87
Product Link http://www.matrixscientific.com/037427.html

Safety Information

hcodes Irritant-Harmful/Stored under Argon/Air Sensitive
MSDS http://www.matrixscientific.com/037427.html
Availability details for
Quantity
  • Availability
  • Add to Cart