MAT196006236 Matrix Scientific

1-(2,4,5-Trimethylphenyl)ethanone

  • MDL number MFCD00017244
  • Cas Number : 2040-07-5
  • Molecular Weight : 162.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 10
Country of Origin USA
Boiling point 246
Product Link http://www.matrixscientific.com/037442.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/037442.html
Availability details for
Quantity
  • Availability
  • Add to Cart