MAT196006308 Matrix Scientific

5-Methyl-1H-pyrazole

  • MDL number MFCD08685900
  • Cas Number : 1453-58-3
  • Molecular Weight : 82.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/037514.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/037514.html
Availability details for
Quantity
  • Availability
  • Add to Cart