MAT196006345 Matrix Scientific

Tricyclo[4.3.1.1~3,8~]undecane-1-carboxylic acid

  • MDL number MFCD00571592
  • Cas Number : 31061-65-1
  • Molecular Weight : 194.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/037551.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/037551.html
Availability details for
Quantity
  • Availability
  • Add to Cart