MAT196006375 Matrix Scientific

1-Benzyl-1H-benzimidazol-2-amine

  • MDL number MFCD00142854
  • Cas Number : 43182-10-1
  • Molecular Weight : 223.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/037581.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/037581.html
Availability details for
Quantity
  • Availability
  • Add to Cart