MAT196006451 Matrix Scientific

8-Methyl-2,3-dihydrofuro[3,2-e]imidazo-[1,2-c]pyrimidine-9-carboxylic acid

  • MDL number MFCD12027146
  • Cas Number : 1092969-14-6
  • Molecular Weight : 219.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/037657.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/037657.html
Availability details for
Quantity
  • Availability
  • Add to Cart