MAT196006471 Matrix Scientific

4-(3-Methyl-1H-indazol-1-yl)-4-oxobutanoic acid

  • MDL number MFCD12027158
  • Cas Number : 902937-57-9
  • Molecular Weight : 232.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/037677.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/037677.html
Availability details for
Quantity
  • Availability
  • Add to Cart