MAT196006501 Matrix Scientific

5-[3-(Trifluoromethyl)phenyl]-1,3-oxazole-4-carboxylic acid

  • MDL number MFCD01934489
  • Cas Number : 253315-30-9
  • Molecular Weight : 257.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/037707.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/037707.html
Availability details for
Quantity
  • Availability
  • Add to Cart