MAT196006504 Matrix Scientific

1-(tert-Butyl) 4-methyl tetrahydro-1,4(2H)-pyridinedicarboxylate

  • MDL number MFCD02183584
  • Cas Number : 124443-68-1
  • Molecular Weight : 243.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 35
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/037710.html

Safety Information

hcodes Irritant/Store in Freezer
MSDS http://www.matrixscientific.com/037710.html
Availability details for
Quantity
  • Availability
  • Add to Cart