MAT196006530 Matrix Scientific

1-(4-Ethoxyphenyl)-4,4,4-trifluorobutane-1,3-dione

  • MDL number MFCD03420708
  • Cas Number : 71712-85-1
  • Molecular Weight : 260.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/037736.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/037736.html
Availability details for
Quantity
  • Availability
  • Add to Cart