MAT196006674 Matrix Scientific

6-Chloro-N,N-dimethyl-1,3,5-triazine-2,4-diamine

  • MDL number MFCD00460497
  • Cas Number : 32998-04-2
  • Molecular Weight : 173.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/037880.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/037880.html
Availability details for
Quantity
  • Availability
  • Add to Cart