MAT196006682 Matrix Scientific

3-(10H-Phenothiazin-10-yl)propanoic acid

  • MDL number MFCD00156933
  • Cas Number : 362-03-8
  • Molecular Weight : 271.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/037888.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/037888.html
Availability details for
Quantity
  • Availability
  • Add to Cart