MAT196006705 Matrix Scientific

(2E)-3-[4-(Diethylamino)phenyl]acrylic acid

  • MDL number MFCD00017303
  • Cas Number : 78776-25-7
  • Molecular Weight : 219.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/037911.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/037911.html
Availability details for
Quantity
  • Availability
  • Add to Cart