MAT196006763 Matrix Scientific

3-[3-(Ethoxycarbonyl)-2-methyl-5-phenyl-1H-pyrrol-1-yl]propanoic acid

  • MDL number MFCD03834505
  • Cas Number : 396123-27-6
  • Molecular Weight : 301.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 131-133
Country of Origin USA
Product Link http://www.matrixscientific.com/037969.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/037969.html
Availability details for
Quantity
  • Availability
  • Add to Cart