MAT196006768 Matrix Scientific

5-(4-Methoxyphenyl)-1,2-dimethyl-1H-pyrrole-3-carboxylic acid

  • MDL number MFCD04154124
  • Cas Number : 724744-85-8
  • Molecular Weight : 245.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 207-209
Country of Origin USA
Product Link http://www.matrixscientific.com/037974.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/037974.html
Availability details for
Quantity
  • Availability
  • Add to Cart