MAT196006789 Matrix Scientific

4-(2-Chloroethyl)-3,5-dimethyl-1H-pyrazole

  • MDL number MFCD00464183
  • Cas Number : 79379-05-8
  • Molecular Weight : 158.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/037995.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/037995.html
Availability details for
Quantity
  • Availability
  • Add to Cart