MAT196006808 Matrix Scientific

3-[3-(Ethoxycarbonyl)-5-(4-fluorophenyl)-2-methyl-1H-pyrrol-1-yl]propanoic acid

  • MDL number MFCD12027260
  • Cas Number : 879329-76-7
  • Molecular Weight : 319.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 107-108
Country of Origin USA
Product Link http://www.matrixscientific.com/038014.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/038014.html
Availability details for
Quantity
  • Availability
  • Add to Cart