MAT196006821 Matrix Scientific

5-(4-Fluorophenyl)-1,2-dimethyl-1H-pyrrole-3-carboxylic acid

  • MDL number MFCD06810919
  • Cas Number : 891764-80-0
  • Molecular Weight : 233.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/038027.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/038027.html
Availability details for
Quantity
  • Availability
  • Add to Cart