MAT196006826 Matrix Scientific

1-[2-(1H-Indol-3-yl)ethyl]-2,5-dimethyl-1H-pyrrole-3-carboxylic acid

  • MDL number MFCD07800638
  • Cas Number : 893772-56-0
  • Molecular Weight : 282.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/038032.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/038032.html
Availability details for
Quantity
  • Availability
  • Add to Cart