MAT196006854 Matrix Scientific

2-(3-Methoxyphenyl)-6-methylimidazo-[1,2-a]pyridine-3-carbaldehyde

  • MDL number MFCD04117112
  • Cas Number : 727975-82-8
  • Molecular Weight : 266.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/038060.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/038060.html
Availability details for
Quantity
  • Availability
  • Add to Cart