MAT196006885 Matrix Scientific

3-[2-(2,5-Dimethyl-1H-pyrrol-1-yl)ethyl]-1H-indole

  • MDL number MFCD02223273
  • Cas Number : 95399-28-3
  • Molecular Weight : 238.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/038091.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/038091.html
Availability details for
Quantity
  • Availability
  • Add to Cart