MAT196006919 Matrix Scientific

Methyl 2-methyl-4,6-dioxocyclohexanecarboxylate

  • MDL number MFCD00154557
  • Cas Number : 56157-27-8
  • Molecular Weight : 184.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/038125.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/038125.html
Availability details for
Quantity
  • Availability
  • Add to Cart