MAT196006971 Matrix Scientific

2-[(tert-Butoxycarbonyl)amino]-3-(1H-imidazol-4-yl)propanoic acid

  • MDL number MFCD00090846
  • Cas Number : 50654-94-9
  • Molecular Weight : 255.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/038177.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/038177.html
Availability details for
Quantity
  • Availability
  • Add to Cart