MAT196007152 Matrix Scientific

Ethyl 1-(2-bromobutanoyl)piperidine-3-carboxylate

  • MDL number MFCD12027307
  • Cas Number : 1119451-39-6
  • Molecular Weight : 306.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/038345.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/038345.html
Availability details for
Quantity
  • Availability
  • Add to Cart