MAT196007179 Matrix Scientific

N-(3-Aminopropyl)-N-butyl-N-methylamine

  • MDL number MFCD06446872
  • Cas Number : 13595-43-2
  • Molecular Weight : 144.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/038372.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/038372.html
Availability details for
Quantity
  • Availability
  • Add to Cart