MAT196007181 Matrix Scientific

N-(3-Chlorophenyl)-N-propylamine

  • MDL number MFCD11144460
  • Cas Number : 31084-60-3
  • Molecular Weight : 169.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/038374.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/038374.html
Availability details for
Quantity
  • Availability
  • Add to Cart